
WIN 62,577

Synonym(s):
17-β-Hydroxy-17-α-ethynyl-δ-4-androstano[3,2-b]pyrimido[1,2-a]benzimidazole
Empirical Formula (Hill Notation):
C29H31N3O
CAS Number:
Molecular Weight:
437.58
Products
Product Category
Brand
Color
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing