Merck
  • [The Study on the Characteristics of Organic Pollution in Typical Herbicide Plant Wastewater].
[The Study on the Characteristics of Organic Pollution in Typical Herbicide Plant Wastewater].
Guang pu xue yu guang pu fen xi = Guang pu (2016-03-12)
Yu-hui Chen, Hong-bo Xi, Dong Yu, Yue-xi Zhou, Xue-min Chen, Xiao-yong Fu
ABSTRACT
Herbicide wastewater is one of tne industrial wastewater, it has high salt content, poor biodegradability, biodegradable characteristics. Nitrogen-containing organic compounds are dominated in dissolved organic matter and dissolved organic matter of wastewater, BOD: COD = 0.045, C:N:P = 692:426:1. Applying static headspace, purg and trap, solid-phase extraction, solid-phase microextraction and liquid-liquid extraction as pretreatment methods combined with gas chromatography/mass spectrometry (GC/MS), which qualitatively analyzed the organic components of the Atrazine, acetochlor herbicide production wastewater and researched the UV spectrum, three-dimensional fluorescence spectroscopy of the wastewater and its major pollutants. The study of GC/MS indicated that Wastewater contained chlorinated hydrocarbons, BTEX and triazines, amides herbicides etc. 38 kinds of volatile and semi-volatile organic compounds, atrazine and acetochlor herbicides accounted for 87. 99%. Affected monocyclic or heterocyclic substances, the ultraviolet absorption spectrum of the wastewater in 210-230 and 250-270 nm in that the amino group lead to the UV absorption red shift 20 nm. Wastewater generated 5 fluorescence peak in λ(ex)/λ(em) = 200-280/300-400 nm, such as a(225/305 nm), b(265/365 nm), c(275/305 nm), d(285/390 nm), e(320/375 nm). Based on three-dimensional fluorescence results of the different functional groups of the characteristics organic, fluorescent area of unsaturated bond is in λ(ex)/λ(em) = 215-230/290-340 nm, the main contribution of the fluorescent substance in the region were olefins, benzene, heterocyclic in the wastewater; fluorescent area of Phenolic hydroxyl and carbonyl is in λ(ex)/λ(em) = 270/300 nm, the main contribution of the fluorescent substance in the region were phenols, ketones.
Product Description
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1, plain gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub