Merck
  • Prolonged and tunable residence time using reversible covalent kinase inhibitors.
Prolonged and tunable residence time using reversible covalent kinase inhibitors.
Nature chemical biology (2015-05-26)
J Michael Bradshaw, Jesse M McFarland, Ville O Paavilainen, Angelina Bisconte, Danny Tam, Vernon T Phan, Sergei Romanov, David Finkle, Jin Shu, Vaishali Patel, Tony Ton, Xiaoyan Li, David G Loughhead, Philip A Nunn, Dane E Karr, Mary E Gerritsen, Jens Oliver Funk, Timothy D Owens, Erik Verner, Ken A Brameld, Ronald J Hill, David M Goldstein, Jack Taunton
ABSTRACT
Drugs with prolonged on-target residence times often show superior efficacy, yet general strategies for optimizing drug-target residence time are lacking. Here we made progress toward this elusive goal by targeting a noncatalytic cysteine in Bruton's tyrosine kinase (BTK) with reversible covalent inhibitors. Using an inverted orientation of the cysteine-reactive cyanoacrylamide electrophile, we identified potent and selective BTK inhibitors that demonstrated biochemical residence times spanning from minutes to 7 d. An inverted cyanoacrylamide with prolonged residence time in vivo remained bound to BTK for more than 18 h after clearance from the circulation. The inverted cyanoacrylamide strategy was further used to discover fibroblast growth factor receptor (FGFR) kinase inhibitors with residence times of several days, demonstrating the generalizability of the approach. Targeting of noncatalytic cysteines with inverted cyanoacrylamides may serve as a broadly applicable platform that facilitates 'residence time by design', the ability to modulate and improve the duration of target engagement in vivo.
Sigma-Aldrich
CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization.