Merck
  • Asymmetric Synthesis of Chiral Secondary Alcohols with Isopropyl Substituents by the Catalytic Enantioselective Addition of Diisopropylzinc to Aldehydes Using N, N-Dialkylnorephedrines as Chiral Catalysts.
Asymmetric Synthesis of Chiral Secondary Alcohols with Isopropyl Substituents by the Catalytic Enantioselective Addition of Diisopropylzinc to Aldehydes Using N, N-Dialkylnorephedrines as Chiral Catalysts.
The Journal of Organic Chemistry (1994)
Soai, Kenso et al.