Merck
  • Juxtacrine Signaling Inhibits Antitumor Immunity by Upregulating PD-L1 Expression.
Juxtacrine Signaling Inhibits Antitumor Immunity by Upregulating PD-L1 Expression.
Cancer research (2018-05-24)
Wen-Hao Yang, Jong-Ho Cha, Weiya Xia, Heng-Huan Lee, Li-Chuan Chan, Ying-Nai Wang, Jennifer L Hsu, Guoxin Ren, Mien-Chie Hung
ABSTRACT
Programmed death-ligand 1 (PD-L1) is a well-known immune checkpoint protein that helps cancer cells evade immune response. Anti-PD-L1 immune therapy has been approved for the treatment of several advanced human cancers. Therefore, further understanding of the regulatory mechanisms of PD-L1 is critical to improve PD-L1-targeting immunotherapy. Recent studies indicated that contact-dependent pathways may regulate anticancer immunity, highlighting the importance of cell contact-induced signaling in cancer immunity. Here, we show that tumor cell contact upregulates PD-L1 expression and reduces T-cell-mediated cell killing through the membrane receptor tyrosine kinase ephrin receptor A10 (EphA10), which is not expressed in normal tissues except testis and is known to mediate cell contact-dependent juxtacrine signaling. Knockout of EphA10 in tumor cells increased T-cell-mediated antitumor immunity in syngeneic mouse models. EphA10 expression also correlated positively with PD-L1 in human breast tumor tissues. Together, our data reveal that in addition to paracrine/autocrine signaling, cell contact-mediated juxtacrine signaling also promotes PD-L1 expression, implying that tumor cells may escape immune surveillance via this mechanism and that targeting EphA10 to boost antitumor immunity may be a new immune checkpoint blockade strategy for female patients with breast cancer.Significance: Regulation of PD-L1 expression by cell contact-mediated signaling promotes immune escape in breast cancer and may lead to the development of an immunotherapy with less adverse effects in female patients. Cancer Res; 78(14); 3761-8. ©2018 AACR.
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade