Merck
  • Conformational stability and catalytic activity of PTEN variants linked to cancers and autism spectrum disorders
Conformational stability and catalytic activity of PTEN variants linked to cancers and autism spectrum disorders
Biochemistry (2015)
Johnston S B and Raines R T