Merck
  • NFATc1-E2F1-LMCD1-Mediated IL-33 Expression by Thrombin Is Required for Injury-Induced Neointima Formation.
NFATc1-E2F1-LMCD1-Mediated IL-33 Expression by Thrombin Is Required for Injury-Induced Neointima Formation.
Arteriosclerosis, thrombosis, and vascular biology (2019-05-03)
Suresh Govatati, Prahalathan Pichavaram, Jagadeesh Janjanam, Baolin Zhang, Nikhlesh K Singh, Arul M Mani, James G Traylor, A Wayne Orr, Gadiparthi N Rao
ABSTRACT
Objective- IL (interleukin)-33 has been shown to play a role in endothelial dysfunction, but its role in atherosclerosis is controversial. Therefore, the purpose of this study is to examine its role in vascular wall remodeling following injury. Approach and Results- Thrombin induced IL-33 expression in a time-dependent manner in human aortic smooth muscle cells and inhibition of its activity by its neutralizing antibody suppressed thrombin induced human aortic smooth muscle cell migration but not DNA synthesis. In exploring the mechanisms, we found that Par1 (protease-activated receptor 1), Gαq/11 (Gα protein q/11), PLCβ3 (phospholipase Cβ3), NFATc1 (nuclear factor of activated T cells), E2F1 (E2F transcription factor 1), and LMCD1 (LIM and cysteine-rich domains protein 1) are involved in thrombin-induced IL-33 expression and migration. Furthermore, we identified an NFAT-binding site at -100 nt that mediates thrombin-induced IL-33 promoter activity. Interestingly, we observed that NFATc1, E2F1, and LMCD1 bind to NFAT site in response to thrombin and found that LMCD1, while alone has no significant effect, enhanced either NFATc1 or E2F1-dependent IL-33 promoter activity. In addition, we found that guidewire injury induces IL-33 expression in SMC and its neutralizing antibodies substantially reduce SMC migration and neointimal growth in vivo. Increased expression of IL-33 was also observed in human atherosclerotic lesions as compared to arteries without any lesions. Conclusions- The above findings reveal for the first time that thrombin-induced human aortic smooth muscle cell migration and injury-induced neointimal growth require IL-33 expression. In addition, thrombin-induced IL-33 expression requires LMCD1 enhanced combinatorial activation of NFATc1 and E2F1.