Merck
  • Suppressive effects of type I angiotensin receptor antagonists, candesartan and irbesartan on allergic asthma.
Suppressive effects of type I angiotensin receptor antagonists, candesartan and irbesartan on allergic asthma.
European journal of pharmacology (2019-02-25)
Mi-Jeong Kim, Dong-Soon Im
ABSTRACT
The effects of candesartan and irbesartan, antagonists of the type I angiotensin II receptor, were investigated on allergic asthma. The antigen-induced degranulation was measured by evaluating β-hexosaminidase activity in vitro. Additionally, a murine ovalbumin-induced allergic asthma model was used to test the in vivo efficacy. It was observed that while candesartan inhibited the antigen-induced degranulation in rat RBL-2H3 mast cells, irbesartan did not. Administration of candesartan and irbesartan decreased the number of immune cells in the bronchoalveolar lavage fluid and reduced the expression of Th2 (IL-4, IL-5, and IL-13) and Th1 cytokines (IL-2 and IFN-γ) in the lung tissues of mice with ovalbumin-induced allergic asthma. Histological studies revealed that both antagonists reduced inflammation and mucin production in the lungs. Therefore, these findings provide evidence that candesartan and irbesartan could have potential applications as anti-allergic agents.
Sigma-Aldrich
Aluminum hydroxide, reagent grade
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
SL327, ≥98% (HPLC), Mixture of E & Z isomers, solid
Sigma-Aldrich
Candesartan cilexetil, ≥98% (HPLC)
Sigma-Aldrich
Irbesartan, ≥98% (HPLC), powder
Sigma-Aldrich
4-Nitrophenyl N-acetyl-β-D-glucosaminide, ≥99% (TLC)
Sigma-Aldrich
Giemsa Stain, Modified Solution, according to Giemsa