Merck
  • Thermal and morphological characterisation of organically modified sepiolite
Thermal and morphological characterisation of organically modified sepiolite
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association (2008)
Tartaglione G, et al.