Merck
  • Hypoosmotic stress induces R loop formation in nucleoli and ATR/ATM-dependent silencing of nucleolar transcription.
Hypoosmotic stress induces R loop formation in nucleoli and ATR/ATM-dependent silencing of nucleolar transcription.
Nucleic acids research (2019-05-23)
Artem K Velichko, Nadezhda V Petrova, Artem V Luzhin, Olga S Strelkova, Natalia Ovsyannikova, Igor I Kireev, Natalia V Petrova, Sergey V Razin, Omar L Kantidze
ABSTRACT
The contribution of nucleoli to the cellular stress response has been discussed for over a decade. Stress-induced inhibition of RNA polymerase I-dependent transcription is hypothesized as a possible effector program in such a response. In this study, we report a new mechanism by which ribosomal DNA transcription can be inhibited in response to cellular stress. Specifically, we demonstrate that mild hypoosmotic stress induces stabilization of R loops in ribosomal genes and thus provokes the nucleoli-specific DNA damage response, which is governed by the ATM- and Rad3-related (ATR) kinase. Activation of ATR in nucleoli strongly depends on Treacle, which is needed for efficient recruitment/retention of TopBP1 in nucleoli. Subsequent ATR-mediated activation of ATM results in repression of nucleolar transcription.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human NBN
Sigma-Aldrich
MISSION® esiRNA, targeting human ATM
Sigma-Aldrich
MISSION® esiRNA, targeting human ATMIN
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-DNA-RNA Hybrid Antibody, clone S9.6, clone S9.6, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human CHD4