Merck
  • Molecular Staples for Tough and Stretchable Adhesion in Integrated Soft Materials.
Molecular Staples for Tough and Stretchable Adhesion in Integrated Soft Materials.
Advanced healthcare materials (2019-08-02)
Baohong Chen, Jiawei Yang, Ruobing Bai, Zhigang Suo
ABSTRACT
The integration of soft materials-biological tissues, gels, and elastomers-is a rapidly developing technology of this time. Whereas hard materials are adhered using adhesives of hard polymers since antiquity, these hard polymers are generally unsuited to adhere soft materials, because hard polymers constrain the deformation of soft materials. This paper describes a design principle to use hard polymers to adhere soft materials, such that adhesion remains tough after the adhered soft materials are subject to many cycles of large stretches in the plane of their interface. The two soft materials have stretchable polymer networks, but need not have functional groups for adhesion. The two soft materials are adhered by forming, in situ at their interface, islands of a hard polymer. The adhesion is tough if the islands themselves are strong, and the polymers of the islands are in topological entanglement with the polymer networks of the soft materials. The adhesion is stretchable if the islands are smaller than the flaw sensitivity length. Several methods of forming the hard polymer islands are demonstrated, and the mechanics and chemistry of adhesion are studied. The design principle will enable many hard polymers to form tough and stretchable adhesion between soft materials.
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%