Merck
  • Reciprocal Regulation of the TOR Kinase and ABA Receptor Balances Plant Growth and Stress Response.
Reciprocal Regulation of the TOR Kinase and ABA Receptor Balances Plant Growth and Stress Response.
Molecular cell (2018-01-02)
Pengcheng Wang, Yang Zhao, Zhongpeng Li, Chuan-Chih Hsu, Xue Liu, Liwen Fu, Yueh-Ju Hou, Yanyan Du, Shaojun Xie, Chunguang Zhang, Jinghui Gao, Minjie Cao, Xiaosan Huang, Yingfang Zhu, Kai Tang, Xingang Wang, W Andy Tao, Yan Xiong, Jian-Kang Zhu
ABSTRACT
As sessile organisms, plants must adapt to variations in the environment. Environmental stress triggers various responses, including growth inhibition, mediated by the plant hormone abscisic acid (ABA). The mechanisms that integrate stress responses with growth are poorly understood. Here, we discovered that the Target of Rapamycin (TOR) kinase phosphorylates PYL ABA receptors at a conserved serine residue to prevent activation of the stress response in unstressed plants. This phosphorylation disrupts PYL association with ABA and with PP2C phosphatase effectors, leading to inactivation of SnRK2 kinases. Under stress, ABA-activated SnRK2s phosphorylate Raptor, a component of the TOR complex, triggering TOR complex dissociation and inhibition. Thus, TOR signaling represses ABA signaling and stress responses in unstressed conditions, whereas ABA signaling represses TOR signaling and growth during times of stress. Plants utilize this conserved phospho-regulatory feedback mechanism to optimize the balance of growth and stress responses.
Sigma-Aldrich
PP242 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse