Merck
  • Manipulation of the dry bean (Phaseolus vulgaris L.) matrix by hydrothermal and high-pressure treatments: Impact on in vitro bile salt-binding ability.
Manipulation of the dry bean (Phaseolus vulgaris L.) matrix by hydrothermal and high-pressure treatments: Impact on in vitro bile salt-binding ability.
Food chemistry (2019-12-08)
Tiantian Lin, Sean O'Keefe, Susan Duncan, Cristina Fernández-Fraguas
ABSTRACT
The capacity of high-fiber foods to sequester BS during digestion is considered a mechanism to lower serum-cholesterol. We investigated the effect of hydrothermal (HT) and high-hydrostatic-pressure (HHP) on the bile salt (BS)-binding ability of dry beans, and how this relates to changes in bean microstructure, fiber content (insoluble-IDF/soluble-SDF), and viscosity. HT and HHP-600 MPa led to significant IDF reduction, including resistant starch (RS), whereas 150-450 MPa significantly increased RS, without modifying IDF/SDF content. Microscopy analysis showed that heating disrupted the bean cell wall integrity, protein matrix and starch granules more severely than 600 MPa; however, tightly-packed complexes of globular starch granules-protein-cell wall fiber formed at HHP ≤ 450 MPa. While HT significantly reduced BS-binding efficiency despite no viscosity change, HHP-treatments maintained or enhanced BS-retention. 600 MPa-treatment induced the maximum BS-binding ability and viscosity. These results demonstrate that BS-binding by beans is not solely based on their fiber content or viscosity, but is influenced by additional microstructural factors.
Product Description
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Taurocholic acid sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications