Merck
  • Epigallocatechin gallate suppresses inflammation in human coronary artery endothelial cells by inhibiting NF-κB.
Epigallocatechin gallate suppresses inflammation in human coronary artery endothelial cells by inhibiting NF-κB.
Life sciences (2020-07-30)
Aravind T Reddy, Sowmya P Lakshmi, E Maruthi Prasad, N Ch Varadacharyulu, Lakshmi Devi Kodidhela
ABSTRACT
The endothelium is a critical regulator of vascular homeostasis, controlling vascular tone and permeability as well as interactions of leukocytes and platelets with blood vessel walls. Consequently, endothelial dysfunction featuring inflammation and reduced vasodilation are considered central to cardiovascular disease (CVD) pathogenesis and have become a therapeutic area of focus. Type II endothelial cell (EC) activation by stress-related stimuli such as tumor necrosis factor-α (TNF-α) initiates the nuclear factor-κB (NF-κB) signaling pathway, a master regulator of inflammatory responses. Because dysregulated NF-κB signaling has been tightly linked to several CVDs, EC-specific inhibition of NF-κB represents an attractive pharmacological strategy. As accumulating evidence highlights the clinical benefits of tea catechin for multiple diseases including CVDs, we sought to determine whether the tea catechin epigallocatechin gallate (EGCG) that displays antioxidative, anti-inflammatory, hypolipidemic, anti-thrombogenic, and anti-hypertensive properties offers protection against CVDs by suppressing the canonical NF-κB pathway. Our findings indicate that EGCG downregulates multiple components of the TNF-α-induced NF-κB signaling pathway and thereby reduces the consequent increase in inflammatory gene transcription and protein expression. Furthermore, EGCG blocked type II EC activation, evidenced by diminished EC leakage and monocyte adhesion in EGCG-treated cells. In summary, our study advances knowledge of EGCG's anti-inflammatory effects on the NF-κB pathway and hence its benefits on endothelial health, supporting its therapeutic potential for CVDs.