- Buffer Standards for the Physiological pH of the Zwitterionic Compound, ACES from 5 to 55°C.
Buffer Standards for the Physiological pH of the Zwitterionic Compound, ACES from 5 to 55°C.
Journal of solution chemistry (2010-02-18)
Rabindra N Roy, Lakshmi N Roy, Michael S Fuge, Chandra N Roy, Curtis A Himes, Paul A Bryant, Kyle T Robinson, Daniel A Kaufmann, Colby H Grove, Toshi Ghosh, Adonis Bwashi
PMID20161482
ABSTRACT
The values of the second dissociation constant p