Merck
Emergence of a High-Plasticity Cell State during Lung Cancer Evolution.
Cancer cell (2020-07-25)
Nemanja Despot Marjanovic, Matan Hofree, Jason E Chan, David Canner, Katherine Wu, Marianna Trakala, Griffin G Hartmann, Olivia C Smith, Jonathan Y Kim, Kelly Victoria Evans, Anna Hudson, Orr Ashenberg, Caroline B M Porter, Alborz Bejnood, Ayshwarya Subramanian, Kenneth Pitter, Yan Yan, Toni Delorey, Devan R Phillips, Nisargbhai Shah, Ojasvi Chaudhary, Alexander Tsankov, Travis Hollmann, Natasha Rekhtman, Pierre P Massion, John T Poirier, Linas Mazutis, Ruifang Li, Joo-Hyeon Lee, Angelika Amon, Charles M Rudin, Tyler Jacks, Aviv Regev, Tuomas Tammela
ABSTRACT
Tumor evolution from a single cell into a malignant, heterogeneous tissue remains poorly understood. Here, we profile single-cell transcriptomes of genetically engineered mouse lung tumors at seven stages, from pre-neoplastic hyperplasia to adenocarcinoma. The diversity of transcriptional states increases over time and is reproducible across tumors and mice. Cancer cells progressively adopt alternate lineage identities, computationally predicted to be mediated through a common transitional, high-plasticity cell state (HPCS). Accordingly, HPCS cells prospectively isolated from mouse tumors and human patient-derived xenografts display high capacity for differentiation and proliferation. The HPCS program is associated with poor survival across human cancers and demonstrates chemoresistance in mice. Our study reveals a central principle underpinning intra-tumoral heterogeneity and motivates therapeutic targeting of the HPCS.
Sigma-Aldrich
Cisplatin, Cisplatin - CAS 15663-27-1, is a platinum coordination complex with potent anti-neoplastic activity. Induces apoptosis in cancer cells, possibly via caspase-3 activation.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-Prosurfactant Protein C (proSP-C) Antibody, serum, Chemicon®