Merck
  • EGFR ligand switch in late stage prostate cancer contributes to changes in cell signaling and bone remodeling.
EGFR ligand switch in late stage prostate cancer contributes to changes in cell signaling and bone remodeling.
The Prostate (2009-01-15)
Alyse M DeHaan, Natalie M Wolters, Evan T Keller, Kathleen M Woods Ignatoski
ABSTRACT
Bone metastasis occurs frequently in advanced prostate cancer (PCa) patients; however, it is not known why this happens. The epidermal growth factor receptor (EGFR) ligand EGF is available to early stage PCa; whereas, TGF-alpha is available when PCa metastasizes. Since the microenvironment of metastases has been shown to play a role in the survival of the tumor, we examined whether the ligands had effects on cell survival and proliferation in early and late PCa. We used LNCaP cells as a model of early stage, non-metastatic PCa and the isogenic C4-2B cells as a model of late stage, metastatic PCa. We found that the proliferation factor MAPK and the survival factor AKT were differentially activated in the presence of different ligands. TGF-alpha induced growth of C4-2B cells and not of the parental LNCaP cells; however, LNCaP cells expressing a constitutively active AKT did proliferate with TGF-alpha. Therefore, AKT appeared to be the TGF-alpha-responsive factor for survival of the late stage PCa cells. LNCaP cells exposed to EGF produced more osteoprotegerin (OPG), an inhibitor of bone remodeling, than C4-2B cells with TGF-alpha, which had increased expression of RANKL, an activator of bone remodeling. In concordance, TGF-alpha-treated C4-2B conditioned medium was able to differentiate an osteoclast precursor line to a greater extent than EGF-treated C4-2B or TGF-alpha-treated LNCaP conditioned media. The switch in EGFR ligand availability as PCa progresses affects cell survival and contributes to bone remodeling.
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, meets USP testing specifications
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material