Merck
  • Selective induction of calcineurin activity and signaling by oligomeric amyloid beta.
Selective induction of calcineurin activity and signaling by oligomeric amyloid beta.
Aging cell (2008-09-11)
Lindsay C Reese, WenRu Zhang, Kelly T Dineley, Rakez Kayed, Giulio Taglialatela
ABSTRACT
Alzheimer's disease (AD) is a terminal age-associated dementia characterized by early synaptic dysfunction and late neurodegeneration. Although the presence of plaques of fibrillar aggregates of the amyloid beta peptide (Abeta) is a signature of AD, evidence suggests that the preplaque small oligomeric Abeta promotes both synaptic dysfunction and neuronal death. We found that young Tg2576 transgenic mice, which accumulate Abeta and develop cognitive impairments prior to plaque deposition, have high central nervous system (CNS) activity of calcineurin (CaN), a phosphatase involved in negative regulation of memory function via inactivation of the transcription factor cAMP responsive element binding proteins (CREB), and display CaN-dependent memory deficits. These results thus suggested the involvement of prefibrillary forms of Abeta. To investigate this issue, we compared the effect of monomeric, oligomeric, and fibrillar Abeta on CaN activity, CaN-dependent pCREB and phosphorylated Bcl-2 Associated death Protein (pBAD) levels, and cell death in SY5Y cells and in rat brain slices, and determined the role of CaN on CREB phosphorylation in the CNS of Tg2576 mice. Our results show that oligomeric Abeta specifically induces CaN activity and promotes CaN-dependent CREB and Bcl-2 Asociated death Protein (BAD) dephosphorylation and cell death. Furthermore, Tg2576 mice display Abeta oligomers and reduced pCREB in the CNS, which is normalized by CaN inhibition. These findings suggest a role for CaN in mediating effects of oligomeric Abeta on neural cells. Because elevated CaN levels have been reported in the CNS of cognitively impaired aged rodents, our results further suggest that abnormal CaN hyperactivity may be a common event exacerbating the cognitive and neurodegenerative impact of oligomeric Abeta in the aging CNS.
Product Description
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture