Merck
  • Effect of Dickkopf-1 and colony stimulating factor-2 on the developmental competence, quality, gene expression and live birth rate of buffalo (Bubalus bubalis) embryos produced by hand-made cloning.
Effect of Dickkopf-1 and colony stimulating factor-2 on the developmental competence, quality, gene expression and live birth rate of buffalo (Bubalus bubalis) embryos produced by hand-made cloning.
Theriogenology (2020-08-22)
S Shyam, P Goel, D Kumar, S Malpotra, M K Singh, S S Lathwal, S Chand, P Palta
ABSTRACT
A functional canonical WNT signaling pathway exists in preimplantation embryos and inhibits embryonic development. Recent studies suggest that this pathway is over-expressed in nuclear transferred (NT), compared to IVF embryos. The present study investigated the effects of Dickkopf-1 (DKK1), an inhibitor of canonical WNT signaling pathway and colony stimulating factor-2 (CSF2), an embryokine, on the developmental competence, quality, gene expression and live birth rate of NT buffalo embryos produced by Hand-made cloning (HMC). Following supplementation of the in vitro culture medium on day 5 with DKK1 (100 ng/mL), CSF2 (10 ng/mL), DKK1+CSF2 or no supplementation (control), the blastocyst rate was higher (P < 0.05) with DKK1 and DKK1+CSF2 (42.6 ± 1.4% and 46.6 ± 0.9%, respectively) than with CSF2 or controls (40.6 ± 1.3% and 39.0 ± 1.3%, respectively). The apoptotic index of the blastocysts was lower (P < 0.05) for DKK1, CSF2 and DKK1+CSF2 groups (3.44 ± 0.14, 3.39 ± 0.11 and 3.11 ± 0.22, respectively) compared to controls (6.64 ± 0.25), and was similar to that of the IVF blastocysts (3.67 ± 0.18). Although the total cell number was similar for the DKK1, CSF2, DKK1+CSF2 and control groups (200.4 ± 3.05, 196.4 ± 3.73, 204.7 ± 3.71 and 205 ± 4.03, respectively), the inner cell mass:trophectoderm cell number ratio of DKK1, CSF2 and DKK1+CSF2 groups (0.21 ± 0.01, 0.17 ± 0.01 and 0.22 ± 0.02, respectively) was higher (P < 0.05) than controls (0.13 ± 0.01) and was similar to that of IVF blastocysts (0.19 ± 0.01). Treatment with DKK1 or CSF2 or both increased (P < 0.05) the expression level of OCT4, NANOG,SOX2, GATA6, BCL2, PTEN, P53, FGF4, GLUT1 and IFN-τ, and decreased that of C-MYC, CDX2, CASPASE, DNMT3a, TCF7 and LEF1 in blastocysts, compared to controls. Transfer of DKK1-treated embryos to 13 recipients resulted in 4 pregnancies (30.8%; 2 live births, one abortion and one currently at 9 months of pregnancy) whereas, transfer of DKK1+CSF2-treated embryos to 16 recipients, resulted in 4 pregnancies (25.0%), all of which resulted in live births. No pregnancy was obtained after transfer of control and CSF-treated embryos to 12 and 16 recipients, respectively. These results suggest that DKK1 treatment of NT embryos increases the blastocyst, conception and live birth rate, and improves their quality whereas, CSF2 treatment, does not affect the blastocyst, conception and live birth rate despite improvement in embryo quality.
Sigma-Aldrich
DKK-1 human, recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture