Merck
  • Synthesis and antibacterial activity of 1-[1,2,4-triazol-3-yl] and 1-[1,3,4-thiadiazol-2-yl]-3-methylthio-6,7-dihydrobenzo[c]thiophen-4(5H)ones.
Synthesis and antibacterial activity of 1-[1,2,4-triazol-3-yl] and 1-[1,3,4-thiadiazol-2-yl]-3-methylthio-6,7-dihydrobenzo[c]thiophen-4(5H)ones.
Bioorganic & medicinal chemistry letters (2005-02-03)
Sara Tehranchian, Tahmineh Akbarzadeh, Mohammad Reza Fazeli, Hossein Jamalifar, Abbas Shafiee
ABSTRACT
A series of 1-[1,2,4-triazol-3-yl] and 1-[1,3,4-thiadiazol-2-yl]-3-methylthio-6,7-dihydrobenzo[c]thiophen-4(5H)ones were synthesized and tested to demonstrate in vitro antimicrobial activity. Some of these compounds exhibited a good activity against Staphylococcus aureus, S. epidermidis and Bacillus subtilis.
Product Description
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV)