Merck
  • The deubiquitinase USP36 Regulates DNA replication stress and confers therapeutic resistance through PrimPol stabilization.
The deubiquitinase USP36 Regulates DNA replication stress and confers therapeutic resistance through PrimPol stabilization.
Nucleic acids research (2020-11-26)
Yuanliang Yan, Zhijie Xu, Jinzhou Huang, Guijie Guo, Ming Gao, Wootae Kim, Xiangyu Zeng, Jake A Kloeber, Qian Zhu, Fei Zhao, Kuntian Luo, Zhenkun Lou
ABSTRACT
PrimPol has been recently identified as a DNA damage tolerant polymerase that plays an important role in replication stress response. However, the regulatory mechanisms of PrimPol are not well defined. In this study, we identify that the deubiquitinase USP36 interferes with degradation of PrimPol to regulate the replication stress response. Mechanistically, USP36 is deubiquitinated following DNA replication stress, which in turn facilitates its upregulation and interaction with PrimPol. USP36 deubiquitinates K29-linked polyubiquitination of PrimPol and increases its protein stability. Depletion of USP36 results in replication stress-related defects and elevates cell sensitivity to DNA-damage agents, such as cisplatin and olaparib. Moreover, USP36 expression positively correlates with the level of PrimPol protein and poor prognosis in patient samples. These findings indicate that the regulation of PrimPol K29-linked ubiquitination by USP36 plays a critical role in DNA replication stress and chemotherapy response.
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human MYC
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution