Merck
A STAT3 palmitoylation cycle promotes TH17 differentiation and colitis.
Nature (2020-10-09)
Mingming Zhang, Lixing Zhou, Yuejie Xu, Min Yang, Yilai Xu, Garrison Paul Komaniecki, Tatsiana Kosciuk, Xiao Chen, Xuan Lu, Xiaoping Zou, Maurine E Linder, Hening Lin
ABSTRACT
Cysteine palmitoylation (S-palmitoylation) is a reversible post-translational modification that is installed by the DHHC family of palmitoyltransferases and is reversed by several acyl protein thioesterases1,2. Although thousands of human proteins are known to undergo S-palmitoylation, how this modification is regulated to modulate specific biological functions is poorly understood. Here we report that the key T helper 17 (TH17) cell differentiation stimulator, STAT33,4, is subject to reversible S-palmitoylation on cysteine 108. DHHC7 palmitoylates STAT3 and promotes its membrane recruitment and phosphorylation. Acyl protein thioesterase 2 (APT2, also known as LYPLA2) depalmitoylates phosphorylated STAT3 (p-STAT3) and enables it to translocate to the nucleus. This palmitoylation-depalmitoylation cycle enhances STAT3 activation and promotes TH17 cell differentiation; perturbation of either palmitoylation or depalmitoylation negatively affects TH17 cell differentiation. Overactivation of TH17 cells is associated with several inflammatory diseases, including inflammatory bowel disease (IBD). In a mouse model, pharmacological inhibition of APT2 or knockout of Zdhhc7-which encodes DHHC7-relieves the symptoms of IBD. Our study reveals not only a potential therapeutic strategy for the treatment of IBD but also a model through which S-palmitoylation regulates cell signalling, which might be broadly applicable for understanding the signalling functions of numerous S-palmitoylation events.
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Red Blood Cell Lysing Buffer Hybri-Max, liquid, sterile-filtered, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Pricing and availability is not currently available.
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Pricing and availability is not currently available.
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Brilliant Blue R, pure
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Pricing and availability is not currently available.