Merck
  • Repression of Irs2 by let-7 miRNAs is essential for homeostasis of the telencephalic neuroepithelium.
Repression of Irs2 by let-7 miRNAs is essential for homeostasis of the telencephalic neuroepithelium.
The EMBO journal (2020-09-29)
Virginia Fernández, Maria Ángeles Martínez-Martínez, Anna Prieto-Colomina, Adrián Cárdenas, Rafael Soler, Martina Dori, Ugo Tomasello, Yuki Nomura, José P López-Atalaya, Federico Calegari, Víctor Borrell
ABSTRACT
Structural integrity and cellular homeostasis of the embryonic stem cell niche are critical for normal tissue development. In the telencephalic neuroepithelium, this is controlled in part by cell adhesion molecules and regulators of progenitor cell lineage, but the specific orchestration of these processes remains unknown. Here, we studied the role of microRNAs in the embryonic telencephalon as key regulators of gene expression. By using the early recombiner Rx-Cre mouse, we identify novel and critical roles of miRNAs in early brain development, demonstrating they are essential to preserve the cellular homeostasis and structural integrity of the telencephalic neuroepithelium. We show that Rx-Cre;DicerF/F mouse embryos have a severe disruption of the telencephalic apical junction belt, followed by invagination of the ventricular surface and formation of hyperproliferative rosettes. Transcriptome analyses and functional experiments in vivo show that these defects result from upregulation of Irs2 upon loss of let-7 miRNAs in an apoptosis-independent manner. Our results reveal an unprecedented relevance of miRNAs in early forebrain development, with potential mechanistic implications in pediatric brain cancer.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-PAX6 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-PAR-3 Antibody, clone 8E8, clone 8E8, from mouse