Merck
  • The NF-κB inhibitor, SC75741, is a novel antiviral against emerging tick-borne bandaviruses.
The NF-κB inhibitor, SC75741, is a novel antiviral against emerging tick-borne bandaviruses.
Antiviral research (2020-12-10)
Crystal A Mendoza, Satoko Yamaoka, Yoshimi Tsuda, Keita Matsuno, Carla M Weisend, Hideki Ebihara
ABSTRACT
Severe fever with thrombocytopenia syndrome virus (SFTSV) and Heartland virus (HRTV) cause viral hemorrhagic fever-like illnesses in humans due to an aberrant host inflammatory response, which contributes to pathogenesis. Here, we established two separate minigenome (MG) systems based on the M-segment of SFTSV and HRTV. Following characterization of both systems for SFTSV and HRTV, we used them as a platform to screen potential compounds that inhibit viral RNA synthesis. We demonstrated that the NF-κB inhibitor, SC75741, reduces viral RNA synthesis of SFTSV and HRTV using our MG platform and validated these results using infectious SFTSV and HRTV. These results may lead to the use of MG systems as potential screening systems for the identification of antiviral compounds and yield novel insights into host-factors that could play role in bandavirus transcription and replication.
Sigma-Aldrich
SC75741, ≥97% (HPLC)
Sigma-Aldrich
Doxycycline hyclate