Merck
  • Cytotoxic 3,5-bis(benzylidene)piperidin-4-ones and N-acyl analogs displaying selective toxicity for malignant cells.
Cytotoxic 3,5-bis(benzylidene)piperidin-4-ones and N-acyl analogs displaying selective toxicity for malignant cells.
European journal of medicinal chemistry (2007-05-15)
Hari N Pati, Umashankar Das, J Wilson Quail, Masami Kawase, Hiroshi Sakagami, Jonathan R Dimmock
ABSTRACT
A series of 3,5-bis(benzylidene)piperidin-4-ones 1, 1-acryloyl-3,5-bis(benzylidene)piperidin-4-ones 2 and adducts of 2 with sodium 2-mercaptoethanesulfonate (mesna), namely series 3, were prepared as candidate cytotoxic agents. These compounds were examined against neoplastic HSC-2, HSC-4 and HL-60 cells as well as HGF, HPC and HPLF normal cell lines and many of the compounds displayed selective toxicity for malignant cells. The CC50 values of the analogs in series 2 towards the cancer cell lines were mainly submicromolar. The relative potencies, selectivity and logP values were in the order of 2>1>3. The sulfonic acid group of a representative compound in series 3 was replaced by a thiol function to produce 4 leading to substantial increases in cytotoxic potencies and hydrophobicity indicating that the presence of a hydrophilic sulfonic acid group was disadvantageous in terms of potency. Molecular modeling suggested that the superior cytotoxicity of various members of series 1-3 over an acyclic analog 5 may have been due to the greater torsion angles theta1 and theta2 created between the arylidene aryl rings and the adjacent olefinic groups in series 1-3.
Product Description
Supelco
Ethylene glycol, analytical standard
Supelco
Sodium 2-mercaptoethanesulfonate, analytical standard, ≥98.0% (titration)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Melphalan, powder