Merck
  • Cardiomyocytes capture stem cell-derived, anti-apoptotic microRNA-214 via clathrin-mediated endocytosis in acute myocardial infarction.
Cardiomyocytes capture stem cell-derived, anti-apoptotic microRNA-214 via clathrin-mediated endocytosis in acute myocardial infarction.
The Journal of biological chemistry (2019-06-21)
Shunsuke Eguchi, Mikito Takefuji, Teruhiro Sakaguchi, Sohta Ishihama, Yu Mori, Takuma Tsuda, Tomonobu Takikawa, Tatsuya Yoshida, Koji Ohashi, Yuuki Shimizu, Ryo Hayashida, Kazuhisa Kondo, Yasuko K Bando, Noriyuki Ouchi, Toyoaki Murohara
ABSTRACT
Extracellular vesicles (EVs) have emerged as key mediators of intercellular communication that have the potential to improve cardiac function when used in cell-based therapy. However, the means by which cardiomyocytes respond to EVs remains unclear. Here, we sought to clarify the role of exosomes in improving cardiac function by investigating the effect of cardiomyocyte endocytosis of exosomes from mesenchymal stem cells on acute myocardial infarction (MI). Exposing cardiomyocytes to the culture supernatant of adipose-derived regenerative cells (ADRCs) prevented cardiomyocyte cell damage under hypoxia in vitro. In vivo, the injection of ADRCs into the heart simultaneous with coronary artery ligation decreased overall cardiac infarct area and prevented cardiac rupture after acute MI. Quantitative RT-PCR-based analysis of the expression of 35 known anti-apoptotic and secreted microRNAs (miRNAs) in ADRCs revealed that ADRCs express several of these miRNAs, among which miR-214 was the most abundant. Of note, miR-214 silencing in ADRCs significantly impaired the anti-apoptotic effects of the ADRC treatment on cardiomyocytes in vitro and in vivo To examine cardiomyocyte endocytosis of exosomes, we cultured the cardiomyocytes with ADRC-derived exosomes labeled with the fluorescent dye PKH67 and found that hypoxic culture conditions increased the levels of the labeled exosomes in cardiomyocytes. Chlorpromazine, an inhibitor of clathrin-mediated endocytosis, significantly suppressed the ADRC-induced decrease of hypoxia-damaged cardiomyocytes and also decreased hypoxia-induced cardiomyocyte capture of both labeled EVs and extracellular miR-214 secreted from ADRCs. Our results indicate that clathrin-mediated endocytosis in cardiomyocytes plays a critical role in their uptake of circulating, exosome-associated miRNAs that inhibit apoptosis.
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder