Merck
  • Gentisyl alcohol derivatives from the marine-derived fungus Penicillium terrestre.
Gentisyl alcohol derivatives from the marine-derived fungus Penicillium terrestre.
Journal of natural products (2008-01-01)
Li Chen, Yuchun Fang, Tianjiao Zhu, Qianqun Gu, Weiming Zhu
ABSTRACT
Nine new gentisyl alcohol derivatives, namely, the trimeric terrestrol A (8), dimeric terrestrols B-H (1-7), and a monomeric derivative (12), together with four known analogues (9-11, 13) were isolated from the marine-derived fungus Penicillium terrestre. The structures of the new compounds were elucidated by spectroscopic methods including one- and two-dimensional NMR as well as low- and high-resolution mass spectrometric analysis. These new compounds (1-8, 12) showed cytotoxic effects on HL-60, MOLT-4, BEL-7402, and A-549 cell lines with IC50 values in the range 5-65 microM. Compound 6 also showed moderate inhibitory activity against protein tyrosine kinases (Src and KDR). Furthermore, all new compounds exhibited moderate radical scavenging activity against DPPH with IC50 values in the range 2.6-8.5 microM.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard