Merck
  • Scavenging of Labile Heme by Hemopexin Is a Key Checkpoint in Cancer Growth and Metastases.
Scavenging of Labile Heme by Hemopexin Is a Key Checkpoint in Cancer Growth and Metastases.
Cell reports (2020-09-24)
Giacomo Canesin, Annalisa Di Ruscio, Mailin Li, Simone Ummarino, Andreas Hedblom, Reeham Choudhury, Agnieszka Krzyzanowska, Eva Csizmadia, Macarena Palominos, Anna Stiehm, Alexander Ebralidze, Shao-Yong Chen, Mahmoud A Bassal, Ping Zhao, Emanuela Tolosano, Laurence Hurley, Anders Bjartell, Daniel G Tenen, Barbara Wegiel
ABSTRACT
Hemopexin (Hx) is a scavenger of labile heme. Herein, we present data defining the role of tumor stroma-expressed Hx in suppressing cancer progression. Labile heme and Hx levels are inversely correlated in the plasma of patients with prostate cancer (PCa). Further, low expression of Hx in PCa biopsies characterizes poorly differentiated tumors and correlates with earlier time to relapse. Significantly, heme promotes tumor growth and metastases in an orthotopic murine model of PCa, with the most aggressive phenotype detected in mice lacking Hx. Mechanistically, labile heme accumulates in the nucleus and modulates specific gene expression via interacting with guanine quadruplex (G4) DNA structures to promote PCa growth. We identify c-MYC as a heme:G4-regulated gene and a major player in heme-driven cancer progression. Collectively, these results reveal that sequestration of labile heme by Hx may block heme-driven tumor growth and metastases, suggesting a potential strategy to prevent and/or arrest cancer dissemination.
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-DNA G-quadruplex (G4) Antibody, clone 1H6, clone 1H6, from mouse