Merck
  • Protocol for the Analysis of Yeast and Human Mitochondrial Respiratory Chain Complexes and Supercomplexes by Blue Native Electrophoresis.
Protocol for the Analysis of Yeast and Human Mitochondrial Respiratory Chain Complexes and Supercomplexes by Blue Native Electrophoresis.
STAR protocols (2020-10-01)
Alba Timón-Gómez, Rafael Pérez-Pérez, Eva Nyvltova, Cristina Ugalde, Flavia Fontanesi, Antoni Barrientos
ABSTRACT
By using negatively charged Coomassie brilliant blue G-250 dye to induce a charge shift on proteins, blue native polyacrylamide gel electrophoresis (BN-PAGE) allows resolution of enzymatically active multiprotein complexes extracted from cellular or subcellular lysates while retaining their native conformation. BN-PAGE was first developed to analyze the size, composition, and relative abundance of the complexes and supercomplexes that form the mitochondrial respiratory chain and OXPHOS system. Here, we present a detailed protocol of BN-PAGE to obtain robust and reproducible results. For complete details on the use and execution of this protocol, please refer to Lobo-Jarne et al. (2018) and Timón-Gómez et al. (2020).
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Nicotinic acid adenine dinucleotide sodium salt, ≥98%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Millipore
Digitonin, High Purity, CAS 11024-24-1 is a non-ionic detergent commonly used to solubilize membrane-bound proteins.
Sigma-Aldrich
Apoferritin from equine spleen
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Digitonin, Used as non-ionic detergent