Merck
  • Guanosine triphosphate links MYC-dependent metabolic and ribosome programs in small-cell lung cancer.
Guanosine triphosphate links MYC-dependent metabolic and ribosome programs in small-cell lung cancer.
The Journal of clinical investigation (2020-10-21)
Fang Huang, Kenneth E Huffman, Zixi Wang, Xun Wang, Kailong Li, Feng Cai, Chendong Yang, Ling Cai, Terry S Shih, Lauren G Zacharias, Andrew Chung, Qian Yang, Milind D Chalishazar, Abbie S Ireland, C Allison Stewart, Kasey Cargill, Luc Girard, Yi Liu, Min Ni, Jian Xu, Xudong Wu, Hao Zhu, Benjamin Drapkin, Lauren A Byers, Trudy G Oliver, Adi F Gazdar, John D Minna, Ralph J DeBerardinis
ABSTRACT
MYC stimulates both metabolism and protein synthesis, but how cells coordinate these complementary programs is unknown. Previous work reported that, in a subset of small-cell lung cancer (SCLC) cell lines, MYC activates guanosine triphosphate (GTP) synthesis and results in sensitivity to inhibitors of the GTP synthesis enzyme inosine monophosphate dehydrogenase (IMPDH). Here, we demonstrated that primary MYChi human SCLC tumors also contained abundant guanosine nucleotides. We also found that elevated MYC in SCLCs with acquired chemoresistance rendered these otherwise recalcitrant tumors dependent on IMPDH. Unexpectedly, our data indicated that IMPDH linked the metabolic and protein synthesis outputs of oncogenic MYC. Coexpression analysis placed IMPDH within the MYC-driven ribosome program, and GTP depletion prevented RNA polymerase I (Pol I) from localizing to ribosomal DNA. Furthermore, the GTPases GPN1 and GPN3 were upregulated by MYC and directed Pol I to ribosomal DNA. Constitutively GTP-bound GPN1/3 mutants mitigated the effect of GTP depletion on Pol I, protecting chemoresistant SCLC cells from IMPDH inhibition. GTP therefore functioned as a metabolic gate tethering MYC-dependent ribosome biogenesis to nucleotide sufficiency through GPN1 and GPN3. IMPDH dependence is a targetable vulnerability in chemoresistant MYChi SCLC.
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saponin, Molecular Biology, used as non-ionic surfactant
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Mizoribine, ≥98% (TLC)
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks