Merck
  • Experimental solubility profiling of marketed CNS drugs, exploring solubility limit of CNS discovery candidate.
Experimental solubility profiling of marketed CNS drugs, exploring solubility limit of CNS discovery candidate.
Bioorganic & medicinal chemistry letters (2010-11-09)
Yun W Alelyunas, James R Empfield, Dennis McCarthy, Russell C Spreen, Khanh Bui, Luciana Pelosi-Kilby, Cindy Shen
ABSTRACT
We determined the experimental solubility of CNS marketed drugs. Of the 98 drugs measured, greater than 90% had solubility >10 μM in pH 7.4 buffer. Only seven drugs had solubility <10 μM. Using these data, we established a solubility criterion to support CNS discovery. The implication of poor solubility with potential safety concerns and undesirable side effects are discussed.
Product Description
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Riluzole, solid