Merck
  • Synthesis of cinnamoyl ketoamides as hybrid structures of antioxidants and calpain inhibitors.
Synthesis of cinnamoyl ketoamides as hybrid structures of antioxidants and calpain inhibitors.
Bioorganic & medicinal chemistry letters (2011-04-21)
Yeong Jae Yoo, Dong Hyuk Nam, Seo Yun Jung, Jae Wan Jang, Hyoung Ja Kim, Changbae Jin, Ae Nim Pae, Yong Sup Lee
ABSTRACT
The excessive calpain activation causes serious cellular damage or even cell death in neurological disorders such as stroke and Alzheimer's disease. Oxidative stress has also been implicated in the initiation or progression of neurodegenerative diseases. In the present studies, a series of cinnamoyl ketoamides 4a-4j were synthesized as hybrid structures of antioxidants and calpain inhibitors. Cinnamoyl ketoamides, possessing an alkyl chain at the α-position, showed potent μ-calpain inhibitory activities indicating that the cinnamoyl skeleton can be regarded as an acyclic variant of calpain inhibitory chromone carboxamide 2. Among synthesized, compound 4e was the most potent inhibitor of μ-calpain (IC(50)=0.13 μM) and also exhibited strong antioxidant activities in DPPH and superoxide anion radical scavenging and lipid peroxidation inhibition assay systems.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)