Merck
  • C1QL3 promotes cell-cell adhesion by mediating complex formation between ADGRB3/BAI3 and neuronal pentraxins.
C1QL3 promotes cell-cell adhesion by mediating complex formation between ADGRB3/BAI3 and neuronal pentraxins.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-12-19)
Matthew J Sticco, Perla A Peña Palomino, David Lukacsovich, Brianna L Thompson, Csaba Földy, Susanne Ressl, David C Martinelli
ABSTRACT
Synapses are the fundamental structural unit by which neurons communicate. An orchestra of proteins regulates diverse synaptic functions, including synapse formation, maintenance, and elimination-synapse homeostasis. Some proteins of the larger C1q super-family are synaptic organizers involved in crucial neuronal processes in various brain regions. C1Q-like (C1QL) proteins bind to the adhesion G protein-coupled receptor B3 (ADGRB3) and act at synapses in a subset of circuits. To investigate the hypothesis that the secreted C1QL proteins mediate tripartite trans-synaptic adhesion complexes, we conducted an in vivo interactome study and identified new binding candidates. We demonstrate that C1QL3 mediates a novel cell-cell adhesion complex involving ADGRB3 and two neuronal pentraxins, NPTX1 and NPTXR. Analysis of single-cell RNA-Seq data from the cerebral cortex shows that C1ql3, Nptx1, and Nptxr are highly co-expressed in the same excitatory neurons. Thus, our results suggest the possibility that in vivo the three co-expressed proteins are presynaptically secreted and form a complex capable of binding to postsynaptically localized ADGRB3, thereby creating a novel trans-synaptic adhesion complex. Identifying new binding partners for C1QL proteins and deciphering their underlying molecular principles will accelerate our understanding of their role in synapse organization.
Sigma-Aldrich
SIGMAFAST BCIP®/NBT, tablet
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)