Merck
  • Antiproliferative triterpenes from the leaves and twigs of Juglans sinensis on HSC-T6 cells.
Antiproliferative triterpenes from the leaves and twigs of Juglans sinensis on HSC-T6 cells.
Journal of natural products (2011-02-12)
Heejung Yang, Eun Ju Jeong, Jinwoong Kim, Sang Hyun Sung, Young Choong Kim
ABSTRACT
Bioassay-guided fractionation of an 80% MeOH extract of leaves and twigs of Juglan sinensis has resulted in the isolation of four new triterpenes (1-4) and 17 known triterpenes (5-21). The new compounds were determined to be 1-oxo-3β,23-dihydroxyolean-12-en-28-oic acid 28-O-β-D-glucopyranoside (1), 1-oxo-3β-hydroxyolean-18-ene (2), 3β,23-dihydroxyurs-12-en-28-oic acid 28-O-β-D-glucopyranoside (3), and 3β,22α-dihydroxyurs-12-en-28-oic acid 28-O-β-D-glucopyranoside (4) by spectroscopic analysis. Compounds 2, 13, 15, and 21 showed antiproliferative activities (14.2, 14.8, 15.6, and 11.0% at 100 μM, respectively) in HSC-T6 cells. Flow cytometry assays revealed that these compounds inhibited HSC-T6 proliferation by inducing apoptosis.
Sigma-Aldrich
Ursolic acid, ≥90%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Supelco
Ursolic acid, analytical standard