Merck
  • Cell-autonomous inflammation of BRCA1-deficient ovarian cancers drives both tumor-intrinsic immunoreactivity and immune resistance via STING.
Cell-autonomous inflammation of BRCA1-deficient ovarian cancers drives both tumor-intrinsic immunoreactivity and immune resistance via STING.
Cell reports (2021-07-22)
Marine Bruand, David Barras, Marco Mina, Eleonora Ghisoni, Matteo Morotti, Evripidis Lanitis, Noémie Fahr, Mathieu Desbuisson, Alizée Grimm, Hualing Zhang, Chloe Chong, Julien Dagher, Sora Chee, Theodora Tsianou, Julien Dorier, Brian J Stevenson, Christian Iseli, Catherine Ronet, Sara Bobisse, Raphael Genolet, Josephine Walton, Michal Bassani-Sternberg, Lana E Kandalaft, Bing Ren, Iain McNeish, Elizabeth Swisher, Alexandre Harari, Mauro Delorenzi, Giovanni Ciriello, Melita Irving, Sylvie Rusakiewicz, Periklis G Foukas, Fabio Martinon, Denarda Dangaj Laniti, George Coukos
ABSTRACT
In this study, we investigate mechanisms leading to inflammation and immunoreactivity in ovarian tumors with homologous recombination deficiency (HRD). BRCA1 loss is found to lead to transcriptional reprogramming in tumor cells and cell-intrinsic inflammation involving type I interferon (IFN) and stimulator of IFN genes (STING). BRCA1-mutated (BRCA1mut) tumors are thus T cell inflamed at baseline. Genetic deletion or methylation of DNA-sensing/IFN genes or CCL5 chemokine is identified as a potential mechanism to attenuate T cell inflammation. Alternatively, in BRCA1mut cancers retaining inflammation, STING upregulates VEGF-A, mediating immune resistance and tumor progression. Tumor-intrinsic STING elimination reduces neoangiogenesis, increases CD8+ T cell infiltration, and reverts therapeutic resistance to dual immune checkpoint blockade (ICB). VEGF-A blockade phenocopies genetic STING loss and synergizes with ICB and/or poly(ADP-ribose) polymerase (PARP) inhibitors to control the outgrowth of Trp53-/-Brca1-/- but not Brca1+/+ ovarian tumors in vivo, offering rational combinatorial therapies for HRD cancers.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-BRCA1 Antibody, clone MS110, clone MS110, from mouse
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein