Merck
  • Design, synthesis, and bio-evaluation of new isoindoline-1,3-dione derivatives as possible inhibitors of acetylcholinesterase.
Design, synthesis, and bio-evaluation of new isoindoline-1,3-dione derivatives as possible inhibitors of acetylcholinesterase.
Research in pharmaceutical sciences (2021-09-16)
Motahareh Hassanzadeh, Farshid Hassanzadeh, Ghadam Ali Khodarahmi, Mahbobe Rostami, Fateme Azimi, Hamid Nadri, Farshad Homayouni Moghadam
ABSTRACT
Alzheimer's disease is considered one of the lead causes of elderly death around the world. A significant decrease in acetylcholine level in the brain is common in most patients with Alzheimer's disease, therefore acetylcholinesterase (AChE) inhibitors such as donepezil and rivastigmine are widely used for patients with limited therapeutic results and major side effects. A series of isoindoline-1,3-dione -N-benzyl pyridinium hybrids were designed, synthesized and evaluated as anti-Alzheimer agents with cholinesterase inhibitory activities. The structure of the compounds were confirmed by various methods of analysis such as HNMR, CNMR, and FT-IR. Molecular modeling studies were also performed to identify the possible interactions between neprilysin and synthesized compounds. The biological screening results indicated that all synthesized compounds displayed potent inhibitory activity with IC50 values ranging from 2.1 to 7.4 μM. Among synthesized compounds, para-fluoro substituted compounds 7a and 7f exhibited the highest inhibitory potency against AChE (IC50 = 2.1 μM). Molecular modeling studies indicated that the most potent compounds were able to interact with both catalytic and peripheral active sites of the enzyme. Also, some of the most potent compounds (7a, 7c, and 7f) demonstrated a neuroprotective effect against H2O2-induced cell death in PC12 neurons. The synthesized compounds demonstrated moderate to good AChE inhibitory effect with results higher than rivastigmine.
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Cell Freezing Medium-DMSO 1×, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
NGF-β from rat, recombinant, expressed in Sf21 cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Horse Serum, Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture