Merck
  • Suppression of breast cancer progression by FBXL16 via oxygen-independent regulation of HIF1α stability.
Suppression of breast cancer progression by FBXL16 via oxygen-independent regulation of HIF1α stability.
Cell reports (2021-11-25)
Yeon-Ju Kim, Yi Zhao, Jae Kyung Myung, Joo Mi Yi, Min-Jung Kim, Su-Jae Lee
ABSTRACT
Triple-negative breast cancers (TNBCs) are characterized by high rates of recurrence and poor clinical outcomes. Deregulated E3 ligases are involved in breast cancer pathogenesis and progression, but the underlying mechanisms are unclear. Here, we find that F-box and leucine-rich repeat protein 16 (FBXL16) acts as a tumor suppressor in TNBCs. FBXL16 directly binds to HIF1α and induces its ubiquitination and degradation, regardless of the tumor microenvironment, resulting in blockade of the HIF1α-mediated epithelial-mesenchymal transition (EMT) and angiogenesis features of breast cancer. In TNBCs, FBXL16 expression is downregulated by the p38/miR-135b-3p axis, and loss of FBXL16 expression restores HIF1α-mediated metastatic features of breast cancer. Low expression of FBXL16 is associated with high-grade and lymph node-positive tumors and poor overall survival of breast cancer. Taken together, these findings demonstrate that modulation of FBXL16 expression may offer a favorable strategy for treatment of patients with metastatic breast cancer, including TNBCs.
Sigma-Aldrich
MG-132, InSolution, ≥98%, 10 mM, reversible proteasome inhibitor
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC50 = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC50 = 34 nM in vitro, 600 nM in cells).
Millipore
JAK Inhibitor I, JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC50 = 15 nM), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC50 = 1 nM).
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC50 = 72 nM) and MEK2 (IC50 = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
JumpStart REDTaq® DNA Polymerase, Hot-start Taq enzyme with inert dye, 10X buffer included