Merck
  • In Vitro Reconstitution of Yeast Translation System Capable of Synthesizing Long Polypeptide and Recapitulating Programmed Ribosome Stalling.
In Vitro Reconstitution of Yeast Translation System Capable of Synthesizing Long Polypeptide and Recapitulating Programmed Ribosome Stalling.
Methods and protocols (2021-07-22)
Riku Nagai, Yichen Xu, Chang Liu, Ayaka Shimabukuro, Nono Takeuchi-Tomita
ABSTRACT
The rates of translation elongation or termination in eukaryotes are modulated through cooperative molecular interactions involving mRNA, the ribosome, aminoacyl- and nascent polypeptidyl-tRNAs, and translation factors. To investigate the molecular mechanisms underlying these processes, we developed an in vitro translation system from yeast, reconstituted with purified translation elongation and termination factors, utilizing CrPV IGR IRES-containing mRNA, which functions in the absence of initiation factors. The system is capable of synthesizing not only short oligopeptides but also long reporter proteins such as nanoluciferase. By setting appropriate translation reaction conditions, such as the Mg2+/polyamine concentration, the arrest of translation elongation by known ribosome-stalling sequences (e.g., polyproline and CGA codon repeats) is properly recapitulated in this system. We describe protocols for the preparation of the system components, manipulation of the system, and detection of the translation products. We also mention critical parameters for setting up the translation reaction conditions. This reconstituted translation system not only facilitates biochemical analyses of translation but is also useful for various applications, such as structural and functional studies with the aim of designing drugs that act on eukaryotic ribosomes, and the development of systems for producing novel functional proteins by incorporating unnatural amino acids by eukaryotic ribosomes.
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
L-Glutamic acid potassium salt monohydrate, ≥99% (HPLC), powder
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Xylene Cyanol FF, BioReagent, Molecular Biology
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial