Merck
  • Microbes exploit death-induced nutrient release by gut epithelial cells.
Microbes exploit death-induced nutrient release by gut epithelial cells.
Nature (2021-08-06)
Christopher J Anderson, Christopher B Medina, Brady J Barron, Laura Karvelyte, Tania Løve Aaes, Irina Lambertz, Justin S A Perry, Parul Mehrotra, Amanda Gonçalves, Kelly Lemeire, Gillian Blancke, Vanessa Andries, Farzaneh Ghazavi, Arne Martens, Geert van Loo, Lars Vereecke, Peter Vandenabeele, Kodi S Ravichandran
ABSTRACT
Regulated cell death is an integral part of life, and has broad effects on organism development and homeostasis1. Malfunctions within the regulated cell death process, including the clearance of dying cells, can manifest in diverse pathologies throughout various tissues including the gastrointestinal tract2. A long appreciated, yet elusively defined relationship exists between cell death and gastrointestinal pathologies with an underlying microbial component3-6, but the direct effect of dying mammalian cells on bacterial growth is unclear. Here we advance a concept that several Enterobacteriaceae, including patient-derived clinical isolates, have an efficient growth strategy to exploit soluble factors that are released from dying gut epithelial cells. Mammalian nutrients released after caspase-3/7-dependent apoptosis boosts the growth of multiple Enterobacteriaceae and is observed using primary mouse colonic tissue, mouse and human cell lines, several apoptotic triggers, and in conventional as well as germ-free mice in vivo. The mammalian cell death nutrients induce a core transcriptional response in pathogenic Salmonella, and we identify the pyruvate formate-lyase-encoding pflB gene as a key driver of bacterial colonization in three contexts: a foodborne infection model, a TNF- and A20-dependent cell death model, and a chemotherapy-induced mucositis model. These findings introduce a new layer to the complex host-pathogen interaction, in which death-induced nutrient release acts as a source of fuel for intestinal bacteria, with implications for gut inflammation and cytotoxic chemotherapy treatment.
Sigma-Aldrich
Spermidine, ≥99% (GC)
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
Sigma-Aldrich
Anti-MLKL Antibody, clone 3H1, clone 3H1, from rat
Sigma-Aldrich
Caspase 3 Colorimetric Activity Assay Kit, DEVD, Caspase-3 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the sequence DEVD.
Sigma-Aldrich
Formate Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
Q-VD-OPh hydrate, ≥95% (HPLC)