Merck
  • Productive Performance and Meat Characteristics of Kids Fed a Red Orange and Lemon Extract.
Productive Performance and Meat Characteristics of Kids Fed a Red Orange and Lemon Extract.
Animals : an open access journal from MDPI (2021-04-04)
Angela Salzano, Sara Damiano, Livia D'Angelo, Gabriele Ballistreri, Salvatore Claps, Domenico Rufrano, Aristide Maggiolino, Gianluca Neglia, Pasquale De Palo, Roberto Ciarcia
ABSTRACT
This study evaluates the animal performance and meat characteristics of 60 Saanen suckling kids daily fed a red orange and lemon extract (RLE), rich in anthocyanins. In our methodology, after colostrum administration, animals are randomly assigned to two treatments: Treatment group (Group RLE; n = 30) that received RLE (90 mg/kg live body weight) as oral food additive, and a control group (Group CON; n = 30) that received a standard diet. Animals are slaughtered after 40 days. The RLE administration did not influence daily weight gain, carcass measurements, or incidences (expressed as a percentage) of different anatomical regions on the whole carcass weight. On the contrary, RLE supplementation significantly improved the oxidative profile of the meat seven days after slaughtering, as demonstrated by the reduced levels of thiobarbituric acid reactive substances (TBARS; p < 0.01) and hydroperoxides (p < 0.01) in Group RLE compared to Group CON. A significant influence of RLE administration is observed on day 7 for yellowness (p < 0.01). There are also lower saturated and higher monounsaturated and polyunsaturated fatty acids concentration in Group RLE meat (p < 0.01), which also shows lower atherogenic and thrombogenic indexes (p < 0.01) compared to Group CON. The study demonstrates that the supplementation of a diet with RLE rich in anthocyanins is effective to improve the meat quality.
Sigma-Aldrich
Lipid Peroxidation (MDA) Assay Kit, sufficient for 100 colorimetric or fluorometric tests
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells