Merck
EphA2 is an essential mediator of UV radiation-induced apoptosis.
Cancer research (2008-03-15)
Guoqi Zhang, Ching-Ni Njauw, Jong Min Park, Chie Naruse, Masahide Asano, Hensin Tsao
ABSTRACT
One of the physiologic consequences of excessive UV radiation (UVR) exposure is apoptosis. This critical response serves to eliminate genetically injured cells and arises, in part, from activation of DNA damage and p53 signaling. Other contributory pathways, however, likely exist but have not been fully characterized. In a recent global screen of UVR response genes in melanocytes, we identified the receptor tyrosine kinase EPHA2. Using a combination of genetic and pharmacologic approaches, we set out to investigate the upstream regulation of EphA2 by UVR and the functional consequences of this effect. We found that the UVR-associated increase in EphA2 occurs in melanocytes, keratinocytes, and fibroblasts from both human and murine sources. More specifically, UVR effectively up-regulated EphA2 individually in p53-null, p63-null, and p73-null murine embryonic fibroblasts (MEF), suggesting that the p53 family of transcription factors is not essential for the observed effect. However, inhibition of mitogen-activated protein kinase (MAPK) signaling by U0126 and PD98059 significantly reduced the UVR response whereas overexpression of oncogenic NRAS led to an increase in EphA2. These results confirm that UVR induces EphA2 by a p53-independent, but MAPK-dependent, mechanism. In response to UV irradiation, Epha2(-/-) MEFs were highly resistant to UVR-mediated cytotoxicity and apoptosis whereas introduction of EphA2 into both wild-type and p53-null MEFs led to activation of an apoptotic program that can be blocked by caspase-8 inhibition. These functional findings suggest that EphA2 is in fact an essential p53-independent, caspase-8-dependent proapoptotic factor induced by UVR.
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC50 = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
Caspase-9 Inhibitor III, The Caspase-9 Inhibitor III, also referenced under CAS 403848-57-7, controls the biological activity of Caspase-9. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Caspase-8 Inhibitor I, Cell-Permeable, The Caspase 8 Inhibitor I, Cell-Permeable controls the biological activity of Caspase-8. This small molecule/inhibitor is primarily used for Cancer applications.