Merck
  • ASPP2 maintains the integrity of mechanically stressed pseudostratified epithelia during morphogenesis.
ASPP2 maintains the integrity of mechanically stressed pseudostratified epithelia during morphogenesis.
Nature communications (2022-02-19)
Christophe Royer, Elizabeth Sandham, Elizabeth Slee, Falk Schneider, Christoffer B Lagerholm, Jonathan Godwin, Nisha Veits, Holly Hathrell, Felix Zhou, Karolis Leonavicius, Jemma Garratt, Tanaya Narendra, Anna Vincent, Celine Jones, Tim Child, Kevin Coward, Chris Graham, Marco Fritzsche, Xin Lu, Shankar Srinivas
ABSTRACT
During development, pseudostratified epithelia undergo large scale morphogenetic events associated with increased mechanical stress. Using a variety of genetic and imaging approaches, we uncover that in the mouse E6.5 epiblast, where apical tension is highest, ASPP2 safeguards tissue integrity. It achieves this by preventing the most apical daughter cells from delaminating apically following division events. In this context, ASPP2 maintains the integrity and organisation of the filamentous actin cytoskeleton at apical junctions. ASPP2 is also essential during gastrulation in the primitive streak, in somites and in the head fold region, suggesting that it is required across a wide range of pseudostratified epithelia during morphogenetic events that are accompanied by intense tissue remodelling. Finally, our study also suggests that the interaction between ASPP2 and PP1 is essential to the tumour suppressor function of ASPP2, which may be particularly relevant in the context of tissues that are subject to increased mechanical stress.
Sigma-Aldrich
Anti-Partitioning-defective 3 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution