Merck
  • Defective repair of topoisomerase I induced chromosomal damage in Huntington's disease.
Defective repair of topoisomerase I induced chromosomal damage in Huntington's disease.
Cellular and molecular life sciences : CMLS (2022-03-01)
Nelma M Palminha, Cleide Dos Santos Souza, Jon Griffin, Chunyan Liao, Laura Ferraiuolo, Sherif F El-Khamisy
ABSTRACT
Topoisomerase1 (TOP1)-mediated chromosomal breaks are endogenous sources of DNA damage that affect neuronal genome stability. Whether TOP1 DNA breaks are sources of genomic instability in Huntington's disease (HD) is unknown. Here, we report defective 53BP1 recruitment in multiple HD cell models, including striatal neurons derived from HD patients. Defective 53BP1 recruitment is due to reduced H2A ubiquitination caused by the limited RNF168 activity. The reduced availability of RNF168 is caused by an increased interaction with p62, a protein involved in selective autophagy. Depletion of p62 or disruption of the interaction between RNAF168 and p62 was sufficient to restore 53BP1 enrichment and subsequent DNA repair in HD models, providing new opportunities for therapeutic interventions. These findings are reminiscent to what was described for p62 accumulation caused by C9orf72 expansion in ALS/FTD and suggest a common mechanism by which protein aggregation perturb DNA repair signaling.
Sigma-Aldrich
Anti-Beta III Tubulin Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Caspase 3 Antibody, active (cleaved) form, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Topoisomerase I-DNA Covalent Complexes Antibody, clone 1.1A, clone 1.1A, from mouse