Merck
  • High-Resolution Optical Tweezers Combined with Multicolor Single-Molecule Microscopy.
High-Resolution Optical Tweezers Combined with Multicolor Single-Molecule Microscopy.
Methods in molecular biology (Clifton, N.J.) (2022-09-06)
Rajeev Yadav, Kasun B Senanayake, Matthew J Comstock
ABSTRACT
We present an instrument that combines high-resolution optical tweezers and multicolor confocal fluorescence spectroscopy. Biological macromolecules exhibit complex conformation and stoichiometry changes in coordination with their motion and activity. To further our understanding of the complex machinery of life, we need methods that can simultaneously probe more than one degree of freedom of single molecules and complexes. Fluorescence optical tweezers, or "fleezers," combine the capabilities of optical tweezers and single-molecule fluorescence microscopy into a single instrument. Here we present the latest generation of a high-resolution fleezers instrument integrated with multicolor fluorescence spectroscopy. The tweezers portion of the instrument can manipulate biological macromolecules with pN scale forces while measuring subnanometer distances. Simultaneous with tweezers measurements, the multicolor fluorescence capability allows the direct observation of multiple molecules or multiple degrees of freedom which allows, for example, the observation of multiple proteins simultaneously within a complex. The instrument incorporates three fluorescence excitation lasers, all sourced from a single-mode optical fiber allowing a reliable alignment scheme, that allows, for example, three independent fluorescent probes or fluorescence resonance energy transfer (FRET) measurements and also increases flexibility in the choice of fluorescent probes. To avoid photobleaching and improve tweezers stability, the instrument implements a timesharing (using a single trap laser to produce a pair of traps via rapid switching between two locations) and interlacing (turning the trapping beam off when the fluorescence excitation beams are on and vice versa) scheme using acousto-optic modulators (AOM) to rapidly and precisely modulate lasers. Our latest "random phase" trap AOM control method obliterates previous residual trap positioning and bead position measurement errors. Here we present the general design principles and detailed construction and testing protocols for the instrument.
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
Pyranose Oxidase from Coriolus sp., recombinant, expressed in E. coli, ≥2.7 units/mg solid