Merck
  • Ribosome changes reprogram translation for chemosurvival in G0 leukemic cells.
Ribosome changes reprogram translation for chemosurvival in G0 leukemic cells.
Science advances (2022-10-29)
Chandreyee Datta, Samuel S Truesdell, Keith Q Wu, Syed I A Bukhari, Harrison Ngue, Brienna Buchanan, Olivier Le Tonqueze, Sooncheol Lee, Swapna Kollu, Madeleine A Granovetter, Myriam Boukhali, Johannes Kreuzer, Maheen S Batool, Leonora Balaj, Wilhelm Haas, Shobha Vasudevan
ABSTRACT
Quiescent leukemic cells survive chemotherapy, with translation changes. Our data reveal that FXR1, a protein amplified in several aggressive cancers, is elevated in quiescent and chemo-treated leukemic cells and promotes chemosurvival. This suggests undiscovered roles for this RNA- and ribosome-associated protein in chemosurvival. We find that FXR1 depletion reduces translation, with altered rRNAs, snoRNAs, and ribosomal proteins (RPs). FXR1 regulates factors that promote transcription and processing of ribosomal genes and snoRNAs. Ribosome changes in FXR1-overexpressing cells, including RPLP0/uL10 levels, activate eIF2α kinases. Accordingly, phospho-eIF2α increases, enabling selective translation of survival and immune regulators in FXR1-overexpressing cells. Overriding these genes or phospho-eIF2α with inhibitors reduces chemosurvival. Thus, elevated FXR1 in quiescent or chemo-treated leukemic cells alters ribosomes that trigger stress signals to redirect translation for chemosurvival.
Sigma-Aldrich
Anti-FXR1 Antibody, clone 6BG10, clone 6BG10, from mouse
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, clone DM1A, Upstate®, from mouse
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®