Merck
Effect of type 2 diabetes on the inducible degrader of LDL receptor.
Journal of lipid research (2023-04-25)
Sum Lam, David Tak Wai Lui, Sammy Wing Ming Shiu, Ying Wong, Kathryn Choon Beng Tan
ABSTRACT
The inducible degrader of LDL receptor (IDOL) acts as a post-transcriptional degrader of the LDL receptor (LDLR). IDOL is functionally active in the liver and in peripheral tissues. We have evaluated IDOL expression in circulating monocytes in subjects with and without type 2 diabetes and determined whether changes in IDOL expression could affect macrophage function like cytokine production in vitro. One hundred forty individuals with type 2 diabetes and 110 healthy control subjects were recruited. Cellular expression of IDOL and LDLR in peripheral blood CD14+ monocytes was measured by flow cytometry. The expression of intracellular IDOL was lower in individuals with diabetes than control (21.3 ± 4.6 mean fluorescence intensity × 1,000 vs. 23.8 ± 6.2, P < 0.01), and this was accompanied by an increase in cell surface LDLR (5.2 ± 3.0 mean fluorescence intensity × 1,000 vs. 4.3 ± 1.5, P < 0.01), LDL binding, and intracellular lipid (P < 0.01). IDOL expression correlated with HbA1c (r = -0.38, P < 0.01) and serum fibroblast growth factor-21 (FGF21) (r = -0.34, P < 0.01). Multivariable regression analysis, including age, sex, BMI, smoking, HbA1c, and log(FGF21), showed that HbA1c and FGF21 were significant independent determinants of IDOL expression. IDOL knockdown human monocyte-derived macrophages produced higher concentrations of interleukin 1 beta, interleukin 6, and TNFα than control macrophages upon stimulation with lipopolysaccharide (all P < 0.01). In conclusion, the expression of IDOL in CD14+ monocytes was decreased in type 2 diabetes and was associated with glycemia and serum FGF21 concentration.