Merck
  • Evaluation of the passage of Lactobacillus gasseri K7 and bifidobacteria from the stomach to intestines using a single reactor model.
Evaluation of the passage of Lactobacillus gasseri K7 and bifidobacteria from the stomach to intestines using a single reactor model.
BMC microbiology (2009-05-12)
Philipp Ritter, Christian Kohler, Ueli von Ah
ABSTRACT
Probiotic bacteria are thought to play an important role in the digestive system and therefore have to survive the passage from stomach to intestines. Recently, a novel approach to simulate the passage from stomach to intestines in a single bioreactor was developed. The advantage of this automated one reactor system was the ability to test the influence of acid, bile salts and pancreatin.Lactobacillus gasseri K7 is a strain isolated from infant faeces with properties making the strain interesting for cheese production. In this study, a single reactor system was used to evaluate the survival of L. gasseri K7 and selected bifidobacteria from our collection through the stomach-intestine passage. Initial screening for acid resistance in acidified culture media showed a low tolerance of Bifidobacterium dentium for this condition indicating low survival in the passage. Similar results were achieved with B. longum subsp. infantis whereas B. animalis subsp. lactis had a high survival.These initial results were confirmed in the bioreactor model of the stomach-intestine passage. B. animalis subsp. lactis had the highest survival rate (10%) attaining approximately 5 x 106 cfu ml-1 compared to the other tested bifidobacteria strains which were reduced by a factor of up to 106. Lactobacillus gasseri K7 was less resistant than B. animalis subsp. lactis but survived at cell concentrations approximately 1000 times higher than other bifidobacteria. In this study, we were able to show that L. gasseri K7 had a high survival rate in the stomach-intestine passage. By comparing the results with a previous study in piglets we could confirm the reliability of our simulation. Of the tested bifidobacteria strains, only B. animalis subsp. lactis showed acceptable survival for a successful passage in the simulation system.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Millipore
Bile bovine, dried, unfractionated