Merck
  • Adhesion of Candida albicans to endothelial cells under physiological conditions of flow.
Adhesion of Candida albicans to endothelial cells under physiological conditions of flow.
Infection and immunity (2009-07-08)
Sarah E W Grubb, Craig Murdoch, Peter E Sudbery, Stephen P Saville, Jose L Lopez-Ribot, Martin H Thornhill
ABSTRACT
Candida albicans is a commensal organism that under certain circumstances can become pathogenic. During systemic infection C. albicans is disseminated via the circulation to distant organs, where it causes multiple organ failure. Despite the severity of systemic C. albicans infection, little is known about the mechanisms involved in the adhesion of this organism to the endothelium lining blood vessels. Previous studies have used static assays to examine adhesion. However, these do not realistically model blood vessels, where circulating C. albicans cells must adhere to the endothelium under conditions of flow and shear stress. Furthermore, there is conflicting evidence concerning the role played by yeast, pseudohyphal, and hyphal forms of C. albicans in adhesion to endothelium. To test the hypothesis that there may be differences in the abilities of these three morphogenic forms of C. albicans to adhere to endothelium under conditions of flow, we developed an in vitro flow adhesion assay. We found that all three forms of C. albicans rapidly bound to confluent endothelial cells under conditions of flow. Maximum adhesion was found at low shear stress levels similar to that found in postcapillary venules. Moreover, yeast forms bound in significantly greater numbers than did pseudohyphal and hyphal forms, respectively, contrasting with previous findings from static assays. These findings are consistent with recent in vivo data suggesting that yeast forms may be capable of adhering to the endothelium and migrating into the tissues before undergoing morphogenic change to cause tissue damage.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, 10 ×, Modified, without calcium, magnesium or sodium bicarbonate
SAFC
Hanks′ Balanced Salt solution, HBSS Modified, with calcium, with magnesium, without phenol red, liquid, suitable for cell culture
SAFC
Hanks′ Balanced Salt solution, HBSS Modified, with phenol red, without calcium, without magnesium, liquid
Sigma-Aldrich
Hanks′ Balanced Salt solution, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture