Merck
  • Standardization of the Papanicolaou stain. I. A comparison of five nuclear stains.
Standardization of the Papanicolaou stain. I. A comparison of five nuclear stains.
Analytical and quantitative cytology and histology (1990-06-01)
E Schulte, D Wittekind
ABSTRACT
The staining characteristics of five nuclear stains used in a Papanicolaou staining procedure were investigated. Alcohol-fixed cervical smears were stained with a modified Papanicolaou procedure using hematoxylin, alcoholic thionin bromide, alcoholic Victoria blue B, gallocyanin or the thionin Feulgen reagent (thionin-SO2) as the nuclear stain. The same anionic counterstain was used for all slides, and the optical densities of cell nuclei and cytoplasm were measured with the IBAS 2000 image analyzer. Alcoholic thionin gave the most intense nuclear stain, with a very high reproducibility of the staining pattern. Hematoxylin showed the highest coefficient of variation of the staining intensity. Both hematoxylin and gallocyanin gave some nonspecific cytoplasmic staining. Thionin-SO2 allowed a quantitative assessment of DNA, but gave a low staining intensity. Staining with the metal complex dyes interfered with subsequent staining with the pararosaniline Feulgen reagent. Alcoholic thioinin is thus recommended as a nuclear stain for cervical cytology in the Papanicolaou procedure, both for image analysis and for visual microscopy.
Sigma-Aldrich
Victoria Blue B, suitable for microscopy
Sigma-Aldrich
Gallocyanine, Dye content 90 %