Merck
  • Trifluoroethanol stabilizes the molten globule state and induces non-amyloidic turbidity in stem bromelain near its isoelectric point.
Trifluoroethanol stabilizes the molten globule state and induces non-amyloidic turbidity in stem bromelain near its isoelectric point.
International journal of biological macromolecules (2011-07-09)
Sandeep Dave, Sahil Mahajan, Vemika Chandra, Pawan Gupta
ABSTRACT
Stem bromelain (SBM) is a therapeutic protein that has been studied for alkaline denaturation in the intestines, the principal site of its absorption. In this study, we investigated fluorinated alcohol 2,2,2-trifluoroethanol (TFE)-induced conformational changes in the specific/pre-molten globule (SMG) state of SBM observed at pH 10 by spectroscopic methods. Far-UV circular dichroism (CD) spectra showed that the protein retained its native-like secondary structure at TFE concentrations of up to 30% with a pronounced minimum at 222 nm, characteristic of a helix. However, addition of slightly higher TFE concentrations (≥40%) resulted in an ∼2.5-fold induction of this helical feature and a time-dependent increase in non-amyloidic turbidity as evidenced by turbidometric, Congo red-binding, and Thioflavin T (ThT)-binding studies. Near-UV CD spectra suggested a gradual but significant loss of tertiary structure at 10-30% TFE. Tryptophan studies showed blue-shifted fluorescence, although the number of accessible tryptophans remained the same up to 30% TFE. The SMG showed enhanced binding of the fluorescent probe 1-anilino-8-naphthalene sulfonic acid (ANS) up to 30% TFE, beyond which binding plateaued. Thermal and guanidine hydrochloride (GdnHCl) transition studies in the near-UV range indicated a single cooperative transition for the SMG state in the presence of 30% TFE, similar to that observed for native SBM at pH 7.0 (although with different T(m)s), unlike the SMG state. TFE (30%) appeared to induce native-like stability to the original SMG. These observations suggest a transformation of the SMG to a characteristic molten globule (MG) conformation at 30% TFE, possibly due to TFE-induced rearrangement of hydrophobic interactions at the protein's isoelectric point.
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, technical, ≥90% (NT)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid
Sigma-Aldrich
Bromelain from pineapple stem, ≥3 units/mg protein
Sigma-Aldrich
Bromelain from pineapple stem, ≥4 units/mg protein, (chromatography purified)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, suitable for fluorescence, ≥95% (TLC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, suitable for fluorescence, ≥95.0% (T)